Learn More
Human Lysosomal Pro-X Carboxypeptidase/PRCP Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB7164-SP
Product Code. 18770648
  • € 201.00 / 25µg
Description
Lysosomal Pro-X Carboxypeptidase/PRCP Monoclonal specifically detects Lysosomal Pro-X Carboxypeptidase/PRCP in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lysosomal Pro-X Carboxypeptidase/PRCP
Monoclonal
LYOPH
Western Blot 1 ug/mL
P42785
PRCP
Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP Met1-His496 Accession # P42785
25 μg
Primary
Detects human Lysosomal Pro-X Carboxypeptidase/PRCP in ELISAs.
Human
Purified
Western Blot
860821
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Angiotensinase C, EC 3.4.16.2, HUMPCP, Lysosomal carboxypeptidase C, lysosomal Pro-X carboxypeptidase, PCPMGC2202, Proline carboxypeptidase, Prolylcarboxypeptidase, prolylcarboxypeptidase (angiotensinase C), prolylcarboxypeptidase isoform 1 preproprotein
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
5547
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.