missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LYST (aa 2848-2948) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65778 (PA5-65778. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be required for sorting endosomal resident proteins into late multivesicular endosomes by a mechanism involving microtubules.
Specifications
Specifications
| Accession Number | Q99698 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1130 |
| Name | Human LYST (aa 2848-2948) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Beige homolog; Chediak-Higashi syndrome 1; CHS; CHS1; lysosomal trafficking regulator; lysosomal-trafficking regulator; LYST |
| Common Name | LYST |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction