missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human M-Cadherin/Cadherin-15 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4096-SP
This item is not returnable.
View return policy
Description
M-Cadherin/Cadherin-15 Polyclonal specifically detects M-Cadherin/Cadherin-15 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| M-Cadherin/Cadherin-15 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cadherin 15, M-cadherin (myotubule), cadherin 15, type 1, M-cadherin (myotubule), Cadherin-14, Cadherin-15, cadherin-3, CDH14, CDH14cadherin-14, CDH15, CDH3cadherin-15, CDHM, MCAD, M-cadherin, MCadherin, MRD3, Muscle cadherin, muscle-cadherin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1013 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P55291 | |
| CDH15 | |
| Mouse myeloma cell line NS0-derived recombinant human M-Cadherin/Cadherin-15 Val22-Ala606 Accession # P55291 | |
| 25 μg | |
| Primary | |
| Detects human M-Cadherin/Cadherin-15 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction