missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human M-CSF Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2161U-100UG
This item is not returnable.
View return policy
Description
M-CSF Monoclonal specifically detects M-CSF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| M-CSF | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human M-CSF. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 26786 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CSF1 | |
| E. coli-derived recombinant human M-CSF | |
| RUO | |
| 1435 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction