missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAMSTR (aa 125-182) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65581 (PA5-65581. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Transcriptional coactivator. Stimulates the transcriptional activity of MEF2C. Stimulates MYOD1 activity in part via MEF2, resulting in an enhancement of skeletal muscle differentiation.
Especificaciones
Especificaciones
| Accession Number | Q6ZN01 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 284358 |
| Name | Human MAMSTR (aa 125-182) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | likely ortholog of MEF2-activating SAP transcriptional regulator; MAMSTR; MASTR; MEF2 activating motif and SAP domain containing transcriptional regulator; MEF2-activating motif and SAP domain-containing transcriptional regulator; MEF2-activating SAP transcriptional regulatory protein |
| Common Name | MAMSTR |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido