missing translation for 'onlineSavingsMsg'
Learn More

Human Matrilin-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18716909
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18716909 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18716909 Supplier R&D Systems Supplier No. MAB7878SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Matrilin-1 Monoclonal specifically detects Matrilin-1 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Matrilin-1
Applications Immunocytochemistry
Classification Monoclonal
Clone 828806
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P21941
Gene Alias cartilage matrix protein, CMP, CRTM, MATN1, matrilin 1, cartilage matrix protein, Matrilin1
Gene Symbols MATN1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Matrilin-1 Ser23-Val496 Accession # P21941
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4146
Test Specificity Detects human Matrilin-1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Matrilin-2, -3, or -4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.