missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Matrilin-3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3017
This item is not returnable.
View return policy
Description
MATN3 Polyclonal antibody specifically detects MATN3 in Human samples. It is validated for Western Blot.
Specifications
| MATN3 | |
| Polyclonal | |
| Biotin | |
| DIPOA, EDM5, HOA, matrilin 3, matrilin-3, Matrilin3, NDUFC2, OADIP, OS2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4148 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NDUFC2 | |
| Mouse myeloma cell line NS0-derived recombinant human Matrilin-3 | |
| 50 μg | |
| Primary | |
| Detects human Matrilin-3 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Matrilin-3 is observed and less than 1% cross-reactivity with recombinant human (rh) Matrilin-2 and rhMatrilin-4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction