missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MCM7 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC9217T-100UG
This item is not returnable.
View return policy
Description
MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| MCM7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CDC47, CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47, P85MCM, PNAS146, PPP1R104, Protein Phosphatase 1 | |
| E. coli-derived recombinant human MCM7 Gly188-Ala328 Accession # P33993 | |
| RUO | |
| 4176 | |
| Human | |
| IgG |
| Flow Cytometry | |
| 2068B | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P33993 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects human MCM7 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction