missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCM8 (aa 116-260) Control Fragment Recombinant Protein

Product Code. 30210874
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210874

Brand: Invitrogen™ RP104804

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65399 (PA5-65399. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein contains the central domain that is conserved among the MCM proteins. This protein has been shown to co-immunoprecipitate with MCM4, 6 and 7, which suggests that it may interact with other MCM proteins and play a role in DNA replication. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UJA3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84515
Name Human MCM8 (aa 116-260) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5730432L01Rik; C20orf154; dJ967N21.5; DNA helicase MCM8; DNA replication licensing factor MCM8; MCM8; MCM8 minichromosome maintenance deficient 8; MGC119522; MGC119523; MGC12866; MGC4816; minichromosome maintenance 8; minichromosome maintenance 8 homologous recombination repair factor; minichromosome maintenance complex component 8; minichromosome maintenance deficient 8; minichromosome maintenance deficient 8 (S. cerevisiae); POF10; REC; REC homolog
Common Name MCM8
Gene Symbol MCM8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERKGSILVDFKELTEGGEVTNLIPDIATELRDAPEKTLACMGLAIHQVLTKDLERHAAELQAQEGLSNDGETMVNVPHIHARVYNYEPLTQLKNVRANYYGKYIALRGTVVRVSNIKPLCTKMAFLCAACGEIQSFPLPDGKYSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.