missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MDM2/HDM2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1244
This item is not returnable.
View return policy
Description
MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human samples. It is validated for Immunoprecipitation.
Specifications
| MDM2/HDM2 | |
| Monoclonal | |
| LYOPH | |
| Immunoprecipitation 2.5 ug/500 ug cell lysate | |
| Q00987 | |
| MDM2 | |
| E. coli-derived recombinant human MDM2/HDM2 Asn3-Pro491 Accession # Q00987 | |
| 100 μg | |
| Primary | |
| Detects human MDM2/HDM2. | |
| Human | |
| Purified |
| Immunoprecipitation | |
| 197702 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4193 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction