missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mesothelin Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB32652N
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry.
Specifications
| Mesothelin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10232 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 420411 | |
| Flow Cytometry 5 uL/10^6 cells | |
| AAH09272 | |
| MSLN | |
| Mouse myeloma cell line NS0-derived recombinant human Mesothelin Glu296-Gly580 Accession # AAH09272 | |
| 100 Tests | |
| Primary | |
| Detects human Mesothelin in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction