missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mesothelin Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 228.00 - € 537.00
Specifications
| Antigen | Mesothelin |
|---|---|
| Clone | 618924 |
| Dilution | Immunocytochemistry 8-25 μg/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714564
![]() |
R&D Systems
MAB11417-100 |
100 μg |
€ 537.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18724444
|
R&D Systems
MAB11417-SP |
25 μg |
€ 228.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for ImmunocytochemistrySpecifications
| Mesothelin | |
| Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
| Mouse myeloma cell line, NS0-derived human Mesothelin, Glu296-Gly580, Accession # Q13421 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 618924 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q13421 | |
| 10232 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title