missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Metabolic Luminex™ Performance Premixed Kit

Product Code. p-200085251
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30001023

Brand: R&D Systems™ FCSTM1906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 6 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.