missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human METTL7A Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
MettL7A Synthetic Peptide, PEP-0755, from Invitrogen.
MettL7A belongs to the methyltransferase superfamily. It is a probable methyltransferase. Methyltransferase is a type of transferase enzyme which transfers a methyl group from a donor to an acceptor. Often methylation occurs on nucleic bases in DNA or amino acids in protein structures. DNA methylation is often utilized to silence and regulate genes without changing the original DNA sequence. DNA methylation may be necessary for normal growth from embryonic stages in mammals. When mutant embryonic stem cells lacking the murine DNA methyltransferase gene were introduced to a germline of mice they caused a recessive lethal phenotype. Methylation may also be linked to cancer development as methylation of tumor suppressor genes promotes tumorgenesis and metastasis.
Specifications
Specifications
| Gene ID (Entrez) | 25840 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9H8H3 |
| Common Name | Human METTL7A |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction