missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mGluR2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4676R-100UG
This item is not returnable.
View return policy
Description
mGluR2 Monoclonal specifically detects mGluR2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| mGluR2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q14416 | |
| GRM2 | |
| Mouse myeloma cell line NS0-derived recombinant human mGluR2 Glu19-Ser498 Accession # Q14416 | |
| RUO | |
| 2912 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 455310 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1B, GPRC1Bmetabotropic glutamate receptor 2, GRM2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog, mGluR7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human mGluR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with the N-terminal regions of recombinant human mGluR1, -3, -4, -5, -7, or -8 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction