missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mGluR3 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB46681V-100UG
This item is not returnable.
View return policy
Description
mGluR3 Monoclonal antibody specifically detects mGluR3 in Human samples. It is validated for Flow Cytometry
Specifications
| mGluR3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| GLUR3, glutamate receptor, metabotropic 3, GPRC1C, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human mGluR3, Asp25-Ser507, Accession # Q14832 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 530510 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| Q14832 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2913 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction