missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MKP2 (aa 333-386) Control Fragment Recombinant Protein

Product Code. 30194690
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194690

Brand: Invitrogen™ RP107115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-66440 (PA5-66440, PA5-145156 (PA5-145156. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DUSP4 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. DUSP4 inactivates ERK1, ERK2 and JNK, is expressed in a variety of tissues, and is localized in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13115
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1846
Name Human MKP2 (aa 333-386) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700078F24Rik; AI844617; BB104621; dual specificity phosphatase 4; dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; DUSP 4; DUSP4; DUSP-4; E130306H24Rik; HVH2; MAP kinase phosphatase 2; mitogen-activated protein kinase phosphatase 2; MKP II; Mkp2; Mkp-2; MKPII; serine/threonine specific protein phosphatase; TYP; VH1 homologous phosphatase 2; VH2
Common Name MKP2
Gene Symbol DUSP4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QVLATSCAAEAASPSGPLRERGKTPATPTSQFVFSFPVSVGVHSAPSSLPYLHS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.