missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-16/MT3-MMP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1785
This item is not returnable.
View return policy
Description
MMP-16/MT3-MMP Monoclonal specifically detects MMP-16/MT3-MMP in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| MMP-16/MT3-MMP | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P51512 | |
| MMP16 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-16/MT3-MMP Ala32-Pro535 Accession # P51512 | |
| 100 μg | |
| Primary | |
| Detects human MMP-16/MT3-MMP in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MMP-14, -15, or -24 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 782005 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chromosome 8 open reading frame 57, DKFZp761D112, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 16 (membrane-inserted), matrix metalloproteinase 16 (membrane-inserted), matrix metalloproteinase-16, Membrane-type matrix metalloproteinase 3, Membrane-type-3 matrix metalloproteinase, MMP16, MMP-16, MMPX2, MMP-X2, MT3MMP, MT3-MMP, MT3-MMPC8orf57, MT-MMP 3, MT-MMP2, MTMMP3, MT-MMP3, Putative transmembrane protein C8orf57 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4325 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction