missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC513T-100UG
This item is not returnable.
View return policy
Description
MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| MMP-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P08254 | |
| MMP3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-3 Tyr18-Cys477 Accession # P08254 | |
| RUO | |
| 4314 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 50647 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP3, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, Stromelysin 1, stromelysin-1, Transin-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects both the pro and active forms of human MMP-3 in direct ELISAs and Western blots. In direct ELISAs, 50-100% cross-reactivity with recombinant mouse MMP-3 is observed, 10% cross-reactivity with recombinant human (rh) MMP-10 is observed and no cross-reactivity with rhMMP-1, -2, -7, -8, -9, -12 or -13 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction