missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-9 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 11 publications
Brand: R&D Systems AF911
This item is not returnable.
View return policy
Description
MMP-9 Polyclonal specifically detects MMP-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| MMP-9 | |
| Polyclonal | |
| Unconjugated | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4318 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| MMP9 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-9 | |
| 100 μg | |
| Primary | |
| Detects human and mouse MMP-9 in Western blots, dot blots, and human MMP-9 by immunocytochemistry. In Western blots, less than 1% cross-reactivity with recombinant human MMP-1, -2, -3, -7, -8, -10, -12, and -13 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction