missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Active Caspase-3 Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 155 publications
Brand: R&D Systems AF835-SP
This item is not returnable.
View return policy
Description
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| Caspase-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 836 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P42574 | |
| CASP3 | |
| KLH-coupled human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # P42574 | |
| 25 μg | |
| Primary | |
| Distinguishes apoptotic cells from non-apoptotic cells in immunohistochemistry. Detects the p17 subunit of Caspase-3, but does not detect the precursor form in Western blot. We recommend R&D Systems Affinity Purified Anti-Caspase-3 (Catalog # AF-605-NA) or Monoclonal Anti-Cleaved-Caspase-3 (Catalog # MAB835) for Western blot applications. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction