missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Alkaline Phosphatase/ALPL Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2909-SP
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPL Polyclonal specifically detects Alkaline Phosphatase/ALPL in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Alkaline Phosphatase/ALPL | |
| Polyclonal | |
| Western Blot 0.5 ug/mL | |
| Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 249 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALPL | |
| E. coli-derived recombinant human Alkaline Phosphatase/ALPL Leu18-Phe524 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Alkaline Phosphatase/ALPL in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction