missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse alpha-Fetoprotein/AFP Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC1368V-100UG
This item is not returnable.
View return policy
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| alpha-Fetoprotein/AFP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| AFP, Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, DSCAM2, FETA, HP, HPAFP | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 174 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 189502 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AFP | |
| Human umbilical cord serum-derived alpha-Fetoprotein/AFP | |
| 100 μg | |
| Primary | |
| Detects human alpha-Fetoprotein/AFP in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only (RUO)
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto