missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Aminopeptidase PILS/ARTS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2334
This item is not returnable.
View return policy
Description
Aminopeptidase PILS/ARTS1 Monoclonal specifically detects Aminopeptidase PILS/ARTS1 in Human, Mouse samples. It is validated for Western Blot, Simple Western.
Specifications
| Aminopeptidase PILS/ARTS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| Q9NZ08 | |
| ERAP1 | |
| S. frugiperda insect ovarian cell line Sf 9-derived recombinant human ARTS1 Accession # Q9NZ08 | |
| 100 μg | |
| Primary | |
| Detects human Aminopeptidase PILS/ARTS1 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse Aminopeptidase PILS/ARTS1 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 6H9 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipocyte-derived leucine aminopeptidase, A-LAP, A-LAPALAP, Aminopeptidase PILS, ARTS1, ARTS1aminopeptidase regulator of TNFR1 shedding, ARTS-1PILSAP, EC 3.4.11, EC 3.4.11.-, EC 3.4.11.1, endoplasmic reticulum aminopeptidase 1, endoplasmic reticulum aminopeptidase associated with antigen processing, ERAAP1, ERAP1, KIAA0525APPILS, PILS-AP, PILS-APERAAP, Puromycin-insensitive leucyl-specific aminopeptidase, Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51752 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction