missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse ATF1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4370-SP
This item is not returnable.
View return policy
Description
ATF1 Polyclonal specifically detects ATF1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| ATF1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activating transcription factor 1FUS/ATF-1, cAMP-dependent transcription factor ATF-1, EWS-ATF1, Protein TREB36, TREB36, TREB36cyclic AMP-dependent transcription factor ATF-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 466 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P18846 | |
| ATF1 | |
| E. coli-derived recombinant human ATF1 Met1-Val271 Accession # P18846 | |
| 25 μg | |
| Primary | |
| Detects human and mouse ATF1 in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction