missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BMP-7 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF354
This item is not returnable.
View return policy
Description
BMP-7 Polyclonal specifically detects BMP-7 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| BMP-7 | |
| Polyclonal | |
| Unconjugated | |
| P18075 | |
| BMP7 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BMP-7 Ser293-His431 Accession # P18075 | |
| 100 μg | |
| Primary | |
| Detects human and mouse BMP-7 in Western blots. In this format, less than 1% cross-reactivity with recombinant human (rh) BMP-2, rhBMP-3, rhBMP-4, rhBMP-5, and rhBMP-6 is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 655 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction