missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Bovine Insulin Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC1417R-100UG
This item is not returnable.
View return policy
Description
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Bovine samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Insulin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| IDDM2, ILPR, INS, insulin, IRDN, MODY10, proinsulin | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects bovine, human, and mouse insulin. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 182410 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| INS | |
| E. coli-derived recombinant human Insulin | |
| RUO | |
| 3630 | |
| Human, Mouse, Bovine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction