missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Carm1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF7277
This item is not returnable.
View return policy
Description
Carm1 Polyclonal specifically detects Carm1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.
Specifications
| Carm1 | |
| Polyclonal | |
| Unconjugated | |
| Q86X55 | |
| CARM1 | |
| E. coli-derived recombinant human Carm1 Lys209-Leu379 Accession # Q86X55 | |
| 100 μg | |
| Primary | |
| Detects human Carm1 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunocytochemistry 5-15 μg/mL, Knockout Validated | |
| coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10498 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction