Learn More
Human/Mouse CD44 Rat, PE-Cy7, Clone: IM7, enQuireBio™
Product Code. 15951938
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15951938 25 μg 25µg
15941938 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15951938 Supplier EnquireBio Supplier No. QAB39PE725ug
Rat Monoclonal Antibody
Specifications
Antigen Human/Mouse CD44
Applications Flow Cytometry
Classification Monoclonal
Clone IM7
Concentration 0.5mg/mL
Conjugate PE-Cyanine7
Formulation 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1, pH7.2
Gene Accession No. NP_001034239
Gene Symbols Cd44
Host Species Rat
Purification Method Protein A/G Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12505
Content And Storage Store at 2-4°C.
Isotype IgG2b κ