missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse COX-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB37401-SP
This item is not returnable.
View return policy
Description
COX-1 Monoclonal specifically detects COX-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| COX-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P23219 | |
| PTGS1 | |
| E.coli-derived recombinant human COX-1 Asn381-Leu599 Accession # P23219 | |
| 25 μg | |
| Primary | |
| Detects human COX-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) COX-4I1 and no cross-reactivity with rhCOX-1 or rhCOX-4I2 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 596002 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COX1, COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS, PTGS1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5742 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction