missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse E-Cadherin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 24 publications
Brand: R&D Systems AF648-SP
This item is not returnable.
View return policy
Description
E-Cadherin Polyclonal specifically detects E-Cadherin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC.
Specifications
| E-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 999 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF, In Situ Hybridization (ISH) | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 0.3-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 0.3-15 ug/mL | |
| P12830 | |
| CDH1 | |
| Mouse myeloma cell line NS0-derived recombinant human E-Cadherin Asp155-Ile707 Accession # P12830 | |
| 25 μg | |
| Primary | |
| Detects human E-Cadherin in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction