missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse EBF-3 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5166
This item is not returnable.
View return policy
Description
EBF-3 Polyclonal specifically detects EBF-3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| EBF-3 | |
| Polyclonal | |
| Unconjugated | |
| O08791-2 | |
| EBF3 | |
| E. coli-derived recombinant mouse EBF-3 Ile399-Ser504 Accession # O08791-2 | |
| 100 μg | |
| Primary | |
| Detects endogenous human and mouse EBF-3 in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| COE3, COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF3, EBF-3, OE-2, Olf-1/EBF-like 2, transcription factor COE3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 253738 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction