missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Gastrokine 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7287-SP
This item is not returnable.
View return policy
Description
Gastrokine 1 Polyclonal specifically detects Gastrokine 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Gastrokine 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 18 kDa antrum mucosa protein, AMP18, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, GKN1, Protein CA11 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 56287 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9CR36 | |
| GKN1 | |
| E. coli-derived recombinant mouse Gastrokine 1 Tyr38-Tyr201 Accession # Q9CR36 | |
| 25 μg | |
| Primary | |
| Detects mouse Gastrokine 1 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Gastrokine 1 is observed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction