missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Ghrelin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB8200-SP
This item is not returnable.
View return policy
Description
Ghrelin Monoclonal specifically detects Ghrelin in Human, Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Ghrelin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, GHRL, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51738 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 883622 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9EQX0 | |
| GHRL | |
| E. coli-derived recombinant mouse Obestatin/Ghrelin prepropeptide Gly24-Lys117 Accession # Q9EQX0 | |
| 25 μg | |
| Primary | |
| Detects mouse and human Ghrelin/Obestatin prepropeptide in ELISA. Detects human Ghrelin peptides (acylated and de-acylated) in ELISA. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction