missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB60851
This item is not returnable.
View return policy
Description
Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Monoclonal specifically detects Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9Y661 | |
| HS3ST4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Gly184-Lys456 Accession # Q9Y661 | |
| 100 μg | |
| Primary | |
| Detects human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in direct ELISAs, and human and mouse HS3ST4 in Western blots. In Western blots, no cross-reactivity with recombinant human HS3ST3B1 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 712011 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 30ST4, 3-OST-4, 3OST4 heparan sulfate 3-O-sulfotransferase-4, EC 2.8.2, EC 2.8.2.23, h3-OST-4, heparan sulfate (glucosamine) 3-O-sulfotransferase 4, Heparan sulfate 3-O-sulfotransferase 4, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4, heparan sulfate glucosamine 3-O-sulfotransferase 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9951 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction