missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB60851-SP
This item is not returnable.
View return policy
Description
Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Monoclonal specifically detects Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9Y661 | |
| HS3ST4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Gly184-Lys456 Accession # Q9Y661 | |
| 25 μg | |
| Primary | |
| Detects human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in direct ELISAs, and human and mouse HS3ST4 in Western blots. In Western blots, no cross-reactivity with recombinant human HS3ST3B1 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 712011 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 30ST4, 3-OST-4, 3OST4 heparan sulfate 3-O-sulfotransferase-4, EC 2.8.2, EC 2.8.2.23, h3-OST-4, heparan sulfate (glucosamine) 3-O-sulfotransferase 4, Heparan sulfate 3-O-sulfotransferase 4, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4, heparan sulfate glucosamine 3-O-sulfotransferase 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9951 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction