missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Histone Deacetylase 8/HDAC8 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4359
This item is not returnable.
View return policy
Description
Histone Deacetylase 8/HDAC8 Polyclonal specifically detects Histone Deacetylase 8/HDAC8 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP).
Specifications
| Histone Deacetylase 8/HDAC8 | |
| Polyclonal | |
| Unconjugated | |
| Q9BY41 | |
| HDAC8 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HDAC8 isoform 1 Met1-Val377 (Leu31Pro) Accession # Q9BY41 | |
| 100 μg | |
| Primary | |
| Detects human HDAC8 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| ChIP Assay | |
| LYOPH | |
| Western Blot 1 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells | |
| EC 3.5.1.98, HD8, HDAC8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55869 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction