missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse HO-1/HMOX1/HSP32 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3776
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
Specifications
| HO-1/HMOX1/HSP32 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| P09601 | |
| HMOX1 | |
| E. coli-derived recombinant human HO-1/HMOX1 Met1-Thr261 Accession # P09601 | |
| 100 μg | |
| Primary | |
| Detects endogenous human and mouse HO-1 in Western blots. In Western blots, this antibody does not cross-react with rhHO-2. | |
| Human, Mouse | |
| Purified |
| Western Blot, KnockDown | |
| 412811 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HMOX1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3162 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction