missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Layilin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2637-SP
This item is not returnable.
View return policy
Description
Layilin Monoclonal specifically detects Layilin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready.
Specifications
| Layilin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NP_849156 | |
| LAYN | |
| Mouse myeloma cell line NS0-derived recombinant human Layilin Ala22-Glu220 Accession # NP_849156 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Layilin in direct ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 328024 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ30977, FLJ31092, layilin, LAYN, LYAN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 143903 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction