missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MBD4 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5935
This item is not returnable.
View return policy
Description
MBD4 Polyclonal specifically detects MBD4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| MBD4 | |
| Polyclonal | |
| Unconjugated | |
| O95243 | |
| MBD4 | |
| E. coli-derived recombinant human MBD4 Met382-Ser580 Accession # O95243 | |
| 100 μg | |
| Primary | |
| Detects human and mouse MBD4 in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8930 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction