missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MYF-5 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4027U-100UG
This item is not returnable.
View return policy
Description
MYF-5 Monoclonal specifically detects MYF-5 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| MYF-5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P13349 | |
| MYF5 | |
| E. coli-derived recombinant human MYF-5 Met1-Leu255 Accession # P13349 | |
| RUO | |
| 4617 | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 593128 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, MYF5, myogenic factor 5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MYF-5 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction