missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Park7/DJ-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3668
This item is not returnable.
View return policy
Description
Park7/DJ-1 Polyclonal specifically detects Park7/DJ-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.
Specifications
| Park7/DJ-1 | |
| Polyclonal | |
| Unconjugated | |
| Q99LX0 | |
| PARK7 | |
| E. coli-derived recombinant mouse Park7/DJ-1 Ala2-Asp189 Accession # Q99LX0 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Park7/DJ-1 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
| DJ1, DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11315 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction