missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Pax3/Pax7 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2457R-100UG
This item is not returnable.
View return policy
Description
Pax3/Pax7 Monoclonal antibody specifically detects Pax3/Pax7 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Pax3/Pax7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_000429 | |
| PAX3 | |
| E. coli-derived recombinant human Pax3 (isoform Pax3a) Met1-Ser215 Accession # NP_000429 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Pax3 and Pax7 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Pax1 is observed, approximately 10% with rhPax6, and less than 2% with rhPax2, rhPax4, rhPax5, and rhPax9. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
| Flow Cytometry | |
| 274212 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5077 | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction