missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse PBEF/Visfatin Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC4044N-100UG
This item is not returnable.
View return policy
Description
PBEF/Visfatin/NAMPT Monoclonal specifically detects PBEF/Visfatin/NAMPT in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| PBEF/Visfatin/NAMPT | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q99KQ4 | |
| NAMPT | |
| Mouse myeloma cell line NS0-derived recombinant mouse PBEF/Visfatin Met1-His491 Accession # Q99KQ4 | |
| RUO | |
| 10135 | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 362616 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 2.4.2.12, MGC117256, NAmPRTase, NAMPT, nicotinamide phosphoribosyltransferase, NMPRTase, PBEF1, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human and mouse PBEF/Visfatin in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human PBEF is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction