Learn More
Human/Mouse PTEN Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems MAB847
Product Code. 18161961
  • ONLINE EXCLUSIVE PRICE
    € 517.00
    € 488.25 / 100µg
    Save € 28.75
    5% Off
Description
PTEN Monoclonal specifically detects PTEN in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTEN
Monoclonal
LYOPH
Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready, Knockout Validated
P60484
PTEN
E. coli-derived recombinant human PTEN Thr2-Val403 Accession # P60484
100 μg
Primary
Detects human and mouse PTEN.
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF, Immunoassay
217702
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5728
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.