missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat 14-3-3 sigma Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4424
This item is not returnable.
View return policy
Description
14-3-3 sigma/Stratifin Polyclonal specifically detects 14-3-3 sigma/Stratifin in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications
| 14-3-3 sigma/Stratifin | |
| Polyclonal | |
| Unconjugated | |
| P31947 | |
| SFN | |
| E. coli-derived recombinant human 14-3-3 sigma Met1-Ser248 Accession # P31947 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat 14-3-3 sigma in Western blots. Does not cross react with other recombinant human 14-3-3 isoforms. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunoassay | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| 14-3-3 protein sigma, 1433 sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, SFN, stratifin, YWHAS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2810 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction