missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Akt2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2315
This item is not returnable.
View return policy
Description
AKT2 Monoclonal specifically detects AKT2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| AKT2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P31751 | |
| AKT2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Akt2 Asn2-Glu481 Accession # P31751 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Akt2. Detects human Akt2 in direct ELISAs and endogenous Akt2 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Akt1 or rhAkt3 is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 302514 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 208 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction