missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat alpha-Internexin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4844
This item is not returnable.
View return policy
Description
alpha-Internexin Polyclonal specifically detects alpha-Internexin in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| alpha-Internexin | |
| Polyclonal | |
| Unconjugated | |
| Q16352 | |
| INA | |
| E. coli-derived recombinant human alpha-Internexin Val230-Glu450 Accession # Q16352 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat alpha-Internexin in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| 66 kDa neurofilament protein, alphaInternexin, alpha-Inx, FLJ18662, FLJ57501, INA, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66, NF-66alpha-internexin, TXBP-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9118 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction