missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat alpha-Smooth Muscle Actin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 59 publications
Brand: R&D Systems MAB1420-SP
This item is not returnable.
View return policy
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC.
Specifications
| alpha-Smooth Muscle Actin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1-0.25 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 0.5-25 ug/mL | |
| AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 59 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, In Situ Hybridization (ISH) | |
| 1A4 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACTA2 | |
| Peptide corresponding to residues EEEDSTALVC of the N-term of human alpha-smooth muscle actin (1) | |
| 25 μg | |
| Primary | |
| Detects human alpha-Smooth Muscle Actin. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction