missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat APBA2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6327-SP
This item is not returnable.
View return policy
Description
APBA2 Polyclonal specifically detects APBA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| APBA2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APBA2 | |
| E. coli-derived recombinant human APBA2 Ala2-Leu165 Accession # Q99767 | |
| 25 μg | |
| Primary | |
| Detects human APBA2 in direct ELISAs and human, mouse and rat in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) APBA1 and rhAPBA3 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q99767 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 321 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction